Assay Details
Gene Symbol: CAPZA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283780.1 | NP_001270709.1 | 1 | 909 | 105 | |
GenBank mRNA | AB168787.1 | - | 1 | 909 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
