Assay Details
Gene Symbol: LOC102128869 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015453273.1 | XP_015308759.1 | 6 - 7 | 743 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
