Assay Details
Gene Symbol: SEC14L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284645.1 | NP_001271574.1 | 8 - 9 | 764 | 77 | |
XM_005567606.2 | XP_005567663.1 | 8 - 9 | 920 | 77 | ||
XM_005567608.2 | XP_005567665.1 | 8 - 9 | 1139 | 77 | ||
XM_005567610.2 | XP_005567667.1 | 7 - 8 | 761 | 77 | ||
GenBank mRNA | AB173616.1 | - | 8 - 9 | 764 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
