Assay Details
| Gene Symbol: TIMM10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005577853.2 | XP_005577910.1 | 2 - 3 | 773 | 75 | |
| XM_005577854.2 | XP_005577911.1 | 2 - 3 | 307 | 75 | ||
More Information
Set Membership: |
|

TIMM10
Genomic Map