Assay Details
Gene Symbol: ZNF516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015439606.1 | XP_015295092.1 | 5 - 6 | 4111 | 120 | |
XM_015439607.1 | XP_015295093.1 | 5 - 6 | 3748 | 120 | ||
XM_015439608.1 | XP_015295094.1 | 5 - 6 | 3717 | 120 | ||
XM_015439609.1 | XP_015295095.1 | 6 - 7 | 4101 | 120 | ||
XM_015439610.1 | XP_015295096.1 | 5 - 6 | 5403 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
