Assay Details
Gene Symbol: WASHC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005572088.2 | XP_005572145.1 | 28 - 29 | 2993 | 150 | |
XM_005572089.2 | XP_005572146.1 | 26 - 27 | 2721 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
