Assay Details
| Gene Symbol: N6AMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005548877.2 | XP_005548934.1 | 5 - 6 | 564 | 138 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

N6AMT1| Gene Symbol: N6AMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005548877.2 | XP_005548934.1 | 5 - 6 | 564 | 138 | |
Set Membership: |
|