Assay Details
| Gene Symbol: PDLIM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556440.2 | XP_005556497.1 | 6 - 7 | 909 | 65 | |
| XM_005556441.2 | XP_005556498.1 | 5 - 6 | 818 | 65 | ||
| XM_005556442.2 | XP_005556499.1 | 5 - 6 | 645 | 65 | ||
More Information
Set Membership: |
|

PDLIM3
Genomic Map