Assay Details
Gene Symbol: TMX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283102.1 | NP_001270031.1 | 3 - 4 | 492 | 60 | |
XM_005568378.2 | XP_005568435.1 | 3 - 4 | 265 | 60 | ||
XM_005568379.2 | XP_005568436.1 | 3 - 4 | 394 | 60 | ||
GenBank mRNA | AB168944.1 | - | 3 - 4 | 494 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
