Assay Details
Gene Symbol: NR0B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005593228.2 | XP_005593285.1 | 1 - 2 | 1393 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
