Assay Details
Gene Symbol: PRR11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005583860.2 | XP_005583917.1 | 5 - 6 | 969 | 77 | |
XM_015437881.1 | XP_015293367.1 | 5 - 6 | 720 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
