Assay Details
| Gene Symbol: BNC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005560292.2 | XP_005560349.1 | 2 - 3 | 265 | 80 | |
| XM_015453188.1 | XP_015308674.1 | 1 - 2 | 209 | 80 | ||
More Information
Set Membership: |
|

BNC1
Genomic Map