Assay Details
| Gene Symbol: TAFA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285077.1 | NP_001272006.1 | 4 - 5 | 821 | 66 | |
| XM_005547566.2 | XP_005547623.1 | 4 - 5 | 646 | 66 | ||
| XM_005547567.2 | XP_005547624.1 | 4 - 5 | 1156 | 66 | ||
| GenBank mRNA | AB172528.1 | - | 4 - 5 | 821 | 66 | |
More Information
Set Membership: |
|

TAFA1
Genomic Map