Assay Details
Gene Symbol: LMTK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015441035.1 | XP_015296521.1 | 8 - 9 | 998 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
