Assay Details
| Gene Symbol: VDAC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005563212.2 | XP_005563269.1 | 3 - 4 | 395 | 83 | |
| XM_005563213.2 | XP_005563270.1 | 3 - 4 | 138 | 83 | ||
More Information
Set Membership: |
|

VDAC3
Genomic Map