Assay Details
| Gene Symbol: DPF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005577294.2 | XP_005577351.1 | 8 - 9 | 947 | 63 | |
| XM_005577295.2 | XP_005577352.1 | 7 - 8 | 904 | 63 | ||
More Information
Set Membership: |
|

DPF2
Genomic Map