Assay Details
Gene Symbol: CACNG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015441049.1 | XP_015296535.1 | 2 - 3 | 412 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
