Assay Details
Gene Symbol: CACNG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015441049.1 | XP_015296535.1 | 3 - 4 | 499 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
