Assay Details
| Gene Symbol: ERO1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005539721.2 | XP_005539778.1 | 8 - 9 | 930 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERO1B| Gene Symbol: ERO1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005539721.2 | XP_005539778.1 | 8 - 9 | 930 | 95 | |
Set Membership: |
|