Assay Details
| Gene Symbol: PGBD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005539510.2 | XP_005539567.1 | 2 - 3 | 1493 | 72 | |
| XM_005539512.2 | XP_005539569.1 | 2 - 3 | 88 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005539510.2, XM_005539512.2 transcripts.
Set Membership: |
|

PGBD2
Genomic Map