Assay Details
Gene Symbol: CCNYL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005574088.2 | XP_005574145.1 | 1 - 2 | 393 | 76 | |
XM_005574089.2 | XP_005574146.1 | 1 - 2 | 393 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
