Assay Details
| Gene Symbol: VSIG8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015455053.1 | XP_015310539.1 | 4 - 5 | 782 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

VSIG8| Gene Symbol: VSIG8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015455053.1 | XP_015310539.1 | 4 - 5 | 782 | 70 | |
Set Membership: |
|