Assay Details
Gene Symbol: ZNF793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005589040.2 | XP_005589097.1 | 5 - 6 | 513 | 98 | |
XM_005589041.2 | XP_005589098.1 | 5 - 6 | 518 | 98 | ||
XM_005589042.2 | XP_005589099.1 | 5 - 6 | 846 | 98 | ||
XM_005589043.2 | XP_005589100.1 | 5 - 6 | 707 | 98 | ||
XM_005589044.2 | XP_005589101.1 | 5 - 6 | 510 | 98 | ||
XM_005589045.2 | XP_005589102.1 | 5 - 6 | 499 | 98 | ||
XM_005589046.2 | XP_005589103.1 | 5 - 6 | 499 | 98 | ||
XM_015441224.1 | XP_015296710.1 | 6 - 7 | 629 | 98 | ||
XM_015441225.1 | XP_015296711.1 | 6 - 7 | 697 | 98 | ||
XM_015441226.1 | XP_015296712.1 | 6 - 7 | 642 | 98 | ||
XM_015441227.1 | XP_015296713.1 | 6 - 7 | 671 | 98 | ||
XM_015441228.1 | XP_015296714.1 | 4 - 5 | 467 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
