Assay Details
| Gene Symbol: HACD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005564737.2 | XP_005564794.1 | 4 - 5 | 559 | 104 | |
| XM_015455614.1 | XP_015311100.1 | 4 - 5 | 627 | 104 | ||
More Information
Set Membership: |
|

HACD1
Genomic Map