Assay Details
| Gene Symbol: TSC22D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005585768.2 | XP_005585825.1 | 2 - 3 | 3505 | 72 | |
| XM_005585771.2 | XP_005585828.1 | 2 - 3 | 470 | 72 | ||
| XM_005585772.2 | XP_005585829.1 | 2 - 3 | 161 | 72 | ||
| XM_005585773.2 | XP_005585830.1 | 2 - 3 | 117 | 72 | ||
| XM_015439241.1 | XP_015294727.1 | 3 - 4 | 3181 | 72 | ||
| GenBank mRNA | AB169919.1 | - | 2 - 3 | 402 | 72 | |
| AB170664.1 | - | 2 - 3 | 392 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005585772.2, XM_005585773.2 transcripts.
Set Membership: |
|

TSC22D1
Genomic Map