Assay Details
| Gene Symbol: RGS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283325.1 | NP_001270254.1 | 1 - 2 | 125 | 66 | |
| GenBank mRNA | AB169604.1 | - | 1 - 2 | 196 | 66 | |
| AB173330.1 | - | 1 - 2 | 125 | 66 | ||
More Information
Set Membership: |
|

RGS5
Genomic Map