Assay Details
Gene Symbol: TMC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285371.1 | NP_001272300.1 | 15 - 16 | 2067 | 68 | |
XM_005591351.2 | XP_005591408.1 | 14 - 15 | 2290 | 68 | ||
XM_015442115.1 | XP_015297601.1 | 15 - 16 | 2241 | 68 | ||
GenBank mRNA | AB168722.1 | - | 15 - 16 | 2067 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
