Assay Details
| Gene Symbol: NR2F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557372.2 | XP_005557429.1 | 1 - 2 | 136 | 163 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005557372.2 transcripts.
Set Membership: |
|

NR2F1
Genomic Map