Assay Details
| Gene Symbol: CYP4V2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556479.2 | XP_005556536.1 | 2 - 3 | 622 | 104 | |
| XM_005556480.2 | XP_005556537.1 | 2 - 3 | 281 | 104 | ||
More Information
Set Membership: |
|

CYP4V2
Genomic Map