Assay Details
| Gene Symbol: UBE2Z | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015438219.1 | XP_015293705.1 | 973 | 68 | ||
| GenBank mRNA | AB171060.1 | - | 2 - 3 | 363 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBE2Z| Gene Symbol: UBE2Z | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015438219.1 | XP_015293705.1 | 973 | 68 | ||
| GenBank mRNA | AB171060.1 | - | 2 - 3 | 363 | 68 | |
Set Membership: |
|