Assay Details
| Gene Symbol: CCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319382.1 | NP_001306311.1 | 1 | 860 | 66 | |
| XM_005546896.2 | XP_005546953.1 | 2 | 1034 | 66 | ||
| XM_005546897.2 | XP_005546954.1 | 2 | 1076 | 66 | ||
| GenBank mRNA | DQ008589.1 | - | 1 | 860 | 66 | |
More Information
Set Membership: |
|

CCR1
Genomic Map