Assay Details
Gene Symbol: PPARG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283099.1 | NP_001270028.1 | 6 - 7 | 765 | 82 | |
XM_015445344.1 | XP_015300830.1 | 4 - 5 | 1151 | 82 | ||
XM_015445345.1 | XP_015300831.1 | 6 - 7 | 982 | 82 | ||
XM_015445346.1 | XP_015300832.1 | 6 - 7 | 970 | 82 | ||
XM_015445347.1 | XP_015300833.1 | 6 - 7 | 2165 | 82 | ||
XM_015445348.1 | XP_015300834.1 | 5 - 6 | 913 | 82 | ||
XM_015445349.1 | XP_015300835.1 | 5 - 6 | 745 | 82 | ||
XM_015445350.1 | XP_015300836.1 | 4 - 5 | 1165 | 82 | ||
GenBank mRNA | AY048694.1 | - | 5 - 6 | 681 | 82 | |
AY048695.1 | - | 4 - 5 | 646 | 82 | ||
AY048696.1 | - | 4 - 5 | 594 | 82 | ||
AY048697.1 | - | 5 - 6 | 909 | 82 | ||
AY048698.1 | - | 5 - 6 | 730 | 82 | ||
AY048699.1 | - | 6 - 7 | 944 | 82 | ||
AY048700.1 | - | 6 - 7 | 765 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
