Assay Details
| Gene Symbol: IL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579633.2 | XP_005579690.1 | 4 - 5 | 1169 | 102 | |
| XM_005579635.2 | XP_005579692.1 | 3 - 4 | 1157 | 102 | ||
| XM_015435711.1 | XP_015291197.1 | 5 - 6 | 1258 | 102 | ||
| XM_015435712.1 | XP_015291198.1 | 4 - 5 | 1189 | 102 | ||
More Information
Set Membership: |
|

IL18
Genomic Map