Assay Details
| Gene Symbol: CCR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005551442.2 | XP_005551499.1 | 3 | 1450 | 86 | |
| XM_015448490.1 | XP_015303976.1 | 4 | 1555 | 86 | ||
More Information
Set Membership: |
|

CCR6
Genomic Map