Assay Details
Gene Symbol: PTGER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005588248.2 | XP_005588305.1 | 1 - 2 | 2225 | 84 | |
XM_005588249.2 | XP_005588306.1 | 2 - 3 | 1302 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
