Assay Details
| Gene Symbol: APOBEC3G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284084.1 | NP_001271013.1 | 3 - 4 | 492 | 92 | |
| XM_005567251.2 | XP_005567308.1 | 4 - 5 | 745 | 92 | ||
| XM_015457182.1 | XP_015312668.1 | 2 - 3 | 374 | 92 | ||
| XM_015457183.1 | XP_015312669.1 | 2 - 3 | 400 | 92 | ||
| GenBank mRNA | AB266488.1 | - | 2 - 3 | 464 | 92 | |
| DC858122.1 | - | 4 - 5 | 675 | 92 | ||
| JX915799.1 | - | 3 - 4 | 494 | 92 | ||
More Information
Set Membership: |
|

APOBEC3G
Genomic Map