Assay Details
| Gene Symbol: SMAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556012.2 | XP_005556069.1 | 5 - 6 | 2977 | 64 | |
| XM_005556013.2 | XP_005556070.1 | 4 - 5 | 1319 | 64 | ||
| XM_015450870.1 | XP_015306356.1 | 4 - 5 | 4349 | 64 | ||
| XM_015450871.1 | XP_015306357.1 | 4 - 5 | 1172 | 64 | ||
| XM_015450872.1 | XP_015306358.1 | 4 - 5 | 972 | 64 | ||
| XM_015450873.1 | XP_015306359.1 | 4 - 5 | 2839 | 64 | ||
| GenBank mRNA | AB174721.1 | - | 3 - 4 | 454 | 64 | |
More Information
Set Membership: |
|

SMAD1
Genomic Map