Assay Details
| Gene Symbol: MAP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557119.2 | XP_005557176.2 | 5 - 6 | 7276 | 62 | |
| XM_015451601.1 | XP_015307087.1 | 4 - 5 | 7005 | 62 | ||
More Information
Set Membership: |
|

MAP1B
Genomic Map