Assay Details
| Gene Symbol: CYP3A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319511.1 | NP_001306440.1 | 7 - 8 | 774 | 140 | |
| XM_015447118.1 | XP_015302604.1 | 7 - 8 | 792 | 140 | ||
| GenBank mRNA | DQ074795.1 | - | 7 - 8 | 774 | 140 | |
More Information
Set Membership: |
|

CYP3A5
Genomic Map