Assay Details
Gene Symbol: TMBIM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283684.1 | NP_001270613.1 | 3 - 4 | 246 | 61 | |
XM_005570786.2 | XP_005570843.1 | 3 - 4 | 209 | 61 | ||
XM_015430633.1 | XP_015286119.1 | 3 - 4 | 355 | 61 | ||
XM_015430634.1 | XP_015286120.1 | 3 - 4 | 417 | 61 | ||
GenBank mRNA | AB169574.1 | - | 3 - 4 | 246 | 61 | |
AB170268.1 | - | 3 - 4 | 240 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
