Assay Details
| Gene Symbol: IL2RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283704.1 | NP_001270633.1 | 3 - 4 | 503 | 59 | |
| GenBank mRNA | JQ288991.1 | - | 3 - 4 | 503 | 59 | |
More Information
Set Membership: |
|

IL2RA
Genomic Map