Assay Details
| Gene Symbol: MACO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544391.2 | XP_005544448.1 | 1 - 2 | 123 | 99 | |
| XM_005544392.2 | XP_005544449.1 | 1 - 2 | 123 | 99 | ||
More Information
Set Membership: |
|

MACO1
Genomic Map