Assay Details
Gene Symbol: TLR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319614.1 | NP_001306543.1 | 2 - 3 | 637 | 140 | |
XM_005556469.2 | XP_005556526.1 | 3 - 4 | 1022 | 140 | ||
XM_015451132.1 | XP_015306618.1 | 5 - 6 | 1344 | 140 | ||
XM_015451133.1 | XP_015306619.1 | 4 - 5 | 1242 | 140 | ||
GenBank mRNA | AB445636.1 | - | 2 - 3 | 637 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
