Assay Details
| Gene Symbol: HSD11B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005540709.2 | XP_005540766.1 | 2 - 3 | 380 | 65 | |
| XM_005540710.2 | XP_005540767.1 | 2 - 3 | 364 | 65 | ||
| GenBank mRNA | AB179465.1 | - | 326 | 65 | ||
More Information
Set Membership: |
|

HSD11B1
Genomic Map