Assay Details
| Gene Symbol: LILRB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015441712.1 | XP_015297198.1 | 7 - 8 | 1093 | 61 | |
| XM_015441713.1 | XP_015297199.1 | 7 - 8 | 1093 | 61 | ||
| XM_015441714.1 | XP_015297200.1 | 7 - 8 | 1093 | 61 | ||
More Information
Set Membership: |
|

LILRB4
Genomic Map