Assay Details
| Gene Symbol: LOC102136135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544397.2 | XP_005544454.1 | 1 - 2 | 499 | 56 | |
| XM_005544398.2 | XP_005544455.1 | 1 - 2 | 491 | 56 | ||
| XM_005544399.2 | XP_005544456.1 | 1 - 2 | 485 | 56 | ||
| XM_005544400.2 | XP_005544457.1 | 1 - 2 | 490 | 56 | ||
| XM_005544401.2 | XP_005544458.1 | 1 - 2 | 494 | 56 | ||
| XM_015440633.1 | XP_015296119.1 | 1 - 2 | 490 | 56 | ||
| XM_015440666.1 | XP_015296152.1 | 1 - 2 | 493 | 56 | ||
| GenBank mRNA | L37054.1 | - | 1 - 2 | 155 | 56 | |
More Information
Set Membership: |
|

LOC102136135
Genomic Map