Assay Details
| Gene Symbol: TUBB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553614.2 | XP_005553671.2 | 4 | 834 | 127 | |
| XM_005553615.2 | XP_005553672.1 | 4 | 1073 | 127 | ||
| GenBank mRNA | AB170262.1 | - | 4 | 936 | 127 | |
More Information
Set Membership: |
|

TUBB
Genomic Map