Assay Details
| Gene Symbol: LOC102136267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543487.2 | XP_005543544.2 | 1 | 65 | 116 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005543487.2 transcripts.
Set Membership: |
|

LOC102136267
Genomic Map