Assay Details
| Gene Symbol: UBE2K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283819.1 | NP_001270748.1 | 4 - 5 | 507 | 116 | |
| XM_015450066.1 | XP_015305552.1 | 4 - 5 | 491 | 116 | ||
| XM_015450067.1 | XP_015305553.1 | 3 - 4 | 485 | 116 | ||
| GenBank mRNA | AB171864.1 | - | 4 - 5 | 507 | 116 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015450066.1, XM_015450067.1 transcripts.
Set Membership: |
|

UBE2K
Genomic Map