Assay Details
Gene Symbol: LOC102139497 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005573990.2 | XP_005574047.1 | 2 - 3 | 740 | 102 | |
XM_015432584.1 | XP_015288070.1 | 2 - 3 | 450 | 102 | ||
XM_015432585.1 | XP_015288071.1 | 3 - 4 | 350 | 102 | ||
XM_015432586.1 | XP_015288072.1 | 3 - 4 | 435 | 102 | ||
XM_015432587.1 | XP_015288073.1 | 2 - 3 | 456 | 102 | ||
XM_015432588.1 | XP_015288074.1 | 3 - 4 | 504 | 102 |
Gene Symbol: LOC102146205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319512.1 | NP_001306441.1 | 162 | 102 | ||
GenBank mRNA | KJ922552.1 | - | 162 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
